Gadolinium ≥99% (REO, rare earth oxide basis), ingot/button, Diameter ∼50.8 mm (2.0 in)

Supplier: Thermo Scientific Chemicals

045106-KF
AA45106-KFEA 339 USD
AA45106-KF
Gadolinium ≥99% (REO, rare earth oxide basis), ingot/button, Diameter ∼50.8 mm (2.0 in)
Gadolinium

Purity: 99% CAS No.: 7440-54-2 Physical Form: ≈175g/piece Appearance: Silver MDL No.: MFCD00011022

Formula: Gd
MW: 157.25
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR