Manganese ≥99.6% (metals basis), powder APS <10 µm
Supplier: Thermo Scientific Chemicals
|
Warning
|
045098-18
045098-30
045098-A1
AA45098-18EA
37.2
USD
AA45098-18
AA45098-30
AA45098-A1
Manganese ≥99.6% (metals basis), powder APS <10 µm
Manganese
Purity: 99.6%
CAS No.: 7439-96-5
UN No.: 3089
MDL No.: MFCD00011111
Formula:
Mn Boiling Pt: 2097 °C (1013 hPa) Melting Pt: 1260 °C Density: 7.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 UN: 3089 ADR: 4.1,III Merck Index: 12,05762 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...