Tin ≥99% (metals basis), shot ≤ 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
045096-22
045096-A1
045096-36
AA45096-22EA
48.6
USD
AA45096-22
AA45096-A1
AA45096-36
Tin ≥99% (metals basis), shot ≤ 3.175 mm (0.125 in)
Tin
Purity: 99%
CAS No.: 7440-31-5
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...