Tin ≥99% (metals basis), shot ≤ 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

045096-22 045096-A1 045096-36
AA45096-22EA 48.6 USD
AA45096-22 AA45096-A1 AA45096-36
Tin ≥99% (metals basis), shot ≤ 3.175 mm (0.125 in)
Tin

Purity: 99% CAS No.: 7440-31-5 Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR