Nickel (II) oxide, green (≥78.5% Ni) Ni 78.5%

Supplier: Thermo Scientific Chemicals
Danger

045094-30 045094-A1
AA45094-30EA 103.03 USD
AA45094-30 AA45094-A1
Nickel (II) oxide, green (≥78.5% Ni) Ni 78.5%
Nickel (II) oxide

CAS No.: 1313-99-1 Molecular Formula: NiO Formula Weight: 74.71 Physical Form: -400 Mesh Powder Density: 6.67 Odor: Odorless Merck Reference: 14,6512 MDL No.: MFCD00011145

Formula: NiO
Melting Pt: 1984 °C
Density: 6.67
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7
Merck Index: 14,6512

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR