Zinc ≥99.9% (metals basis), foil, Thickness 0.01+/-0.0025 mm (0.0004+/-0.0001 in)

Supplier: Thermo Scientific Chemicals

045059-FF 045059-FI
AA45059-FFEA 279 USD
AA45059-FF AA45059-FI
Zinc ≥99.9% (metals basis), foil, Thickness 0.01+/-0.0025 mm (0.0004+/-0.0001 in)
Zinc

Purity: 99.9% CAS No.: 7440-66-6 Physical Form: ≈0.045g/25x25mm Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR