Sulfuric acid standardised solution 0.05 N in aqueous solution

Supplier: Thermo Scientific Chemicals

045048-K2 045048-K7
AA45048-K2EA 66.82 USD
AA45048-K2 AA45048-K7 AA45048-M4
Sulfuric acid standardised solution 0.05 N in aqueous solution
Sulfuric acid

Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Odor: Odorless MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR