Tetramethylammonium hydroxide 2,38% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade

Supplier: Thermo Scientific Chemicals
Danger

044940-AK 044940-K2 044940-LV 044940-LU
AA44940-AKEA 43.4 USD
AA44940-AK AA44940-K2 AA44940-LV AA44940-LU
Tetramethylammonium hydroxide 2,38% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade
Tetramethylammonium hydroxide

Purity: 99.9999% CAS No.: 75-59-2 Molecular Formula: C4H13NO Formula Weight: 91.15 Physical Form: Liquid Sensitivity: Air Sensitive Appearance: Colorless to pale yellow Beilstein Reference: 3558708 Merck Reference: 14,9224 UN No.: 1835 MDL No.: MFCD00008280

Formula: C₄H₁₃NO
MW: 91.15
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 III
Merck Index: 14,09224

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR