Silver conductive coating

Supplier: Thermo Scientific Chemicals
Danger

044937-10
AA44937-10EA 202 USD
AA44937-10
Silver conductive coating
Silver (gold coated)

Sheet resistance 0.015 ohms/square in./mil (25um).


Physical Form: Sheet resistance 0.015 ohms/square in./mil (25um). UN No.: 1133 MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 56 °C
Density: 1.62 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
UN: 1133
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR