Chromium etchant
Supplier: Thermo Scientific Chemicals
|
Danger
|
Applications: High purity ceric ammonium nitrate system for precise, clean etching of chromium and chromium oxide films
Physical Form: Liquid
Density: 1.16
Appearance: Orange
Odor: Acrid
UN No.: 3093
MDL No.: MFCD07370678
Applications: High purity ceric ammonium nitrate system for precise, clean etching of chromium and chromium oxide films
Density:
1.16 Storage Temperature: Ambient |
MDL Number:
MFCD07370678 UN: 3093 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...