Nickel etchant for electrodeposited nickel films

Supplier: Thermo Scientific Chemicals
Danger

044580-R0
AA44580-R0EA 96.9 USD
AA44580-R0
Nickel etchant for electrodeposited nickel films
Nickel etchant

Applications: High purity etchant for electrodeposited nickel films


Physical Form: Liquid UN No.: 1760 MDL No.: MFCD09039236


Applications: High purity etchant for electrodeposited nickel films

Storage Temperature: Ambient
MDL Number: MFCD09039236
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR