Chromium ≥99.99% (metals basis), powder -60 mesh

Supplier: Thermo Scientific Chemicals

044556-22 044556-36
AA44556-22EA 158 USD
AA44556-22 AA44556-36
Chromium ≥99.99% (metals basis), powder -60 mesh
Chromium

Purity: 99.99% CAS No.: 7440-47-3 Odor: Odorless MDL No.: MFCD00010944

Formula: Cr
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR