Copper ≥99.999% (metals basis), wire, Puratronic®, Diameter 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals

044550-G1 044550-G5
AA44550-G9EA 1711.86 USD
AA44550-G9 AA44550-G1 AA44550-G5
Copper ≥99.999% (metals basis), wire, Puratronic®, Diameter 1.5 mm (0.06 in)
Copper

Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈15.8g/m Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR