Aluminium ≥99.99% (metals basis), foil, annealed, Thickness 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals

044491-HP 044491-FI 044491-GH
AA44491-HPEA 700 USD
AA44491-HP AA44491-FI AA44491-GH
Aluminium ≥99.99% (metals basis), foil, annealed, Thickness 1.5 mm (0.06 in)
Aluminium

Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈10.125g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR