Gallium metal ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

044418-22 044418-36
AA44418-22EA 496 USD
AA44418-22 AA44418-36
Gallium metal ≥99.999% (metals basis)
Gallium
Formula: Ga
Boiling Pt: 2403 °C (1013 hPa)
Melting Pt: 29 °C
Density: 5.904 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III
Merck Index: 12,04364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR