Poly(styrene) atactic, M.W. 125,000-250,000

Supplier: Thermo Scientific Chemicals

044416-09 044416-18
AA44416-09EA 30.4 USD
AA44416-09 AA44416-18
Poly(styrene) atactic, M.W. 125,000-250,000
Poly(styrene)

CAS No.: 9003-53-6 Formula Weight: 800-5000 Physical Form: Flakes Odor: Odorless MDL No.: MFCD00084450

Formula: C₈H₈
MW: 125000 - 250000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084450
CAS Number: 9003-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR