Platinum 5% in graphite Type 286

Supplier: Thermo Scientific Chemicals
Warning

044365-06 044365-22 044365-14
AA44365-06EA 82.29 USD
AA44365-06 AA44365-22 AA44365-14
Platinum 5% in graphite Type 286
Platinum

Uses: Material for gas sensors


CAS No.: 7440-06-4 Physical Form: Powder Appearance: Dark grey Odor: Odorless UN No.: 1325 MDL No.: MFCD00011179


Applications: Material for gas sensors

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR