Lithium sulfate, anhydrous ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
044352-14
044352-22
AA44352-14EA
137
USD
AA44352-14
AA44352-22
AA44352-36
Lithium sulfate, anhydrous ≥99.99% (metals basis)
Lithium sulphate
MDL: MFCD00011086
Purity: 99.99%
CAS No.: 10377-48-7
Molecular Formula: Li2SO4
Formula Weight: 109.94
Physical Form: Coarse Powder
Density: 2.22
Appearance: White
Odor: Odorless
Melting Point: 845°
Merck Reference: 14,5541
MDL No.: MFCD00011086
Formula:
Li₂O₄S Boiling Pt: 1377 °C (1013 hPa) Melting Pt: 845 °C Density: 2.22 Storage Temperature: Ambient |
MDL Number:
MFCD00011086 CAS Number: 10377-48-7 EINECS: 233-820-4 Merck Index: 14,5541 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...