Aluminium ≥99.9998% (metals basis), wire, Puratronic®, annealed, Diameter 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

044335-G1 044335-G5
AA44335-G1EA 148.18 USD
AA44335-G1 AA44335-G5
Aluminium ≥99.9998% (metals basis), wire, Puratronic®, annealed, Diameter 2.0 mm (0.08 in)
Aluminium

MDL: MFCD00134029


Purity: 99.9998% CAS No.: 7429-90-5 Physical Form: ≈8.4g/m, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR