Aluminium ≥99.9999% (metals basis), foil, Puratronic®, annealed, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

044331-FI 044331-GH 044331-HP
AA44331-FIEA 77.8 USD
AA44331-FI AA44331-GH AA44331-HP
Aluminium ≥99.9999% (metals basis), foil, Puratronic®, annealed, Thickness 0.25 mm (0.01 in)
Aluminium

MDL: MFCD00134029


Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈1.7g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR