Gallium ≥99.99999% (metals basis), pellets, Diameter 6 mm

Supplier: Thermo Scientific Chemicals
Warning

044325-09 044325-18
AA44325-09EA 126 USD
AA44325-09 AA44325-18
Gallium ≥99.99999% (metals basis), pellets, Diameter 6 mm
Gallium

MDL: MFCD00134045


Purity: 99.99999% CAS No.: 7440-55-3 Molecular Formula: Requires overnight shipping with cold pack Storage Temperature: 2°C to 8°C Physical Form: max. weight 0.7g per pellet Appearance: Silver grey UN No.: 2803 MDL No.: MFCD00134045

Formula: Ga
Boiling Pt: 2403 °C (1013 hPa)
Melting Pt: 29 °C
Density: 5.904 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III
Merck Index: 12,04364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR