Gallium ≥99.9999% (metals basis), pellets

Supplier: Thermo Scientific Chemicals
Warning

044324-09 044324-18
AA44324-09EA 148.65 USD
AA44324-09 AA44324-18
Gallium ≥99.9999% (metals basis), pellets
Gallium

Purity: 99.9999% CAS No.: 7440-55-3 Storage Temperature: 2°C to 8°C Physical Form: 0.7 grams/pellet maximum Appearance: Silver grey UN No.: 2803 MDL No.: MFCD00134045

Formula: Ga
Boiling Pt: 2403 °C (1013 hPa)
Melting Pt: 29 °C
Density: 5.904 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III
Merck Index: 12,04364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR