Gallium ≥99.9999% (metals basis), pellets
Supplier: Thermo Scientific Chemicals
|
Warning
|
044324-09
044324-18
AA44324-09EA
148.65
USD
AA44324-09
AA44324-18
Gallium ≥99.9999% (metals basis), pellets
Gallium
Purity: 99.9999%
CAS No.: 7440-55-3
Storage Temperature: 2°C to 8°C
Physical Form: 0.7 grams/pellet maximum
Appearance: Silver grey
UN No.: 2803
MDL No.: MFCD00134045
Formula:
Ga Boiling Pt: 2403 °C (1013 hPa) Melting Pt: 29 °C Density: 5.904 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00134045 CAS Number: 7440-55-3 EINECS: 231-163-8 UN: 2803 ADR: 8,III Merck Index: 12,04364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...