Boron (≥96% B-11) (metals basis), crystalline powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

044257-06
AA44257-06EA 104198.96 USD
AA44257-06
Boron (≥96% B-11) (metals basis), crystalline powder -100 mesh
Boron

MDL: MFCD00134034 Notes: Neutron reflector


Purity: 99.9% CAS No.: 7440-42-8 Physical Form: Powder Odor: Odorless UN No.: 3178 MDL No.: MFCD00134034

Formula: B
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1,III
Merck Index: 13,01333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR