Titanium ≥99.6% (metals basis), thin foil, Thickness 0.0114 mm (0.00045 in)

Supplier: Thermo Scientific Chemicals
Warning

044242-GH 044242-GW
AA44242-GHEA 240 USD
AA44242-GH AA44242-GW
Titanium ≥99.6% (metals basis), thin foil, Thickness 0.0114 mm (0.00045 in)
Titanium

MDL: MFCD00011264


Purity: 99.6% CAS No.: 7440-32-6 Molecular Formula: Commercial Pure Grade 4 Physical Form: ≈0.514g/100x100mm, Light Tight Odor: Odorless

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR