Lithium perchlorate, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
044225-09
044225-30
044225-18
AA44225-09EA
70.9
USD
AA44225-09
AA44225-30
AA44225-18
Lithium perchlorate, anhydrous 99%
Lithium perchlorate
Lithium battery
Purity: 99%
CAS No.: 7791-03-9
Molecular Formula: ClLiO4
Formula Weight: 106.40
Physical Form: Granular, Ampouled under argon
Density: 2.43
Appearance: White
Melting Point: 236°
Boiling Point: 400° dec.
Merck Reference: 14,5539
UN No.: 1481
MDL No.: MFCD00011079
Lithium battery
Formula:
ClLiO₄ Boiling Pt: 400 °C (dec.) Melting Pt: 236 °C Density: 2.43 Storage Temperature: Ambient |
MDL Number:
MFCD00011079 CAS Number: 7791-03-9 EINECS: 232-237-2 UN: 1481 ADR: 5.1,II Merck Index: 14,5539 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...