Dow Corning® High-Vacuum Grease, Thermo Scientific
Supplier: Thermo Scientific Chemicals
044224-KT
AA44224-KTEA 119.24 USD
AA44224-KT
Dow Corning® High-Vacuum Grease, Thermo Scientific
Lubricants
High vacuum grease in a 5.3 oz tube (150 g)
  • Odorless
Applications: Recommended for vacuums reaching between 1 x 10-5 and 1 x 10-6 mm Hg