Docusate sodium ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Dioctyl sulfosuccinate sodium salt, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulfonate, Sodium dioctyl sulfosuccinate, Dioctyl sulfosuccinate, sodium salt, bis(2-Ethylhexyl) sodium sulfosuccinate
Wetting and solubilizing agent
Purity: 96%
CAS No.: 577-11-7
Molecular Formula: C20H37NaO7S
Formula Weight: 444.55
Physical Form: Solid
Density: 1.1
Melting Point: 153-157°
Solubility: Soluble in water, acetone, alcohol
Merck Reference: 14,3401
MDL No.: MFCD00012455
Wetting and solubilizing agent
Formula:
C₂₀H₃₇NaO₇S MW: 444.57 g/mol Boiling Pt: ∼180 °C (1013 hPa) Melting Pt: 155 Density: 1.1 Storage Temperature: Ambient |
MDL Number:
MFCD00012455 CAS Number: 577-11-7 EINECS: 209-406-4 Merck Index: 14,3401 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...