Zinc chloride, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

044198-06 044198-14 044198-22
AA44198-06EA 98.93 USD
AA44198-06 AA44198-14 AA44198-22
Zinc chloride, ultra dry ≥99.999% (metals basis)
Zinc chloride

Purity: 99.999% CAS No.: 7646-85-7 Molecular Formula: ZnCl2 Formula Weight: 136.29 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.91 Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295

Formula: ZnCl₂
Boiling Pt: 732 °C
Melting Pt: 290 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR