Platinum ≥99.9% (metals basis), wire, Diameter 0.01 mm (0.0004 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

044196-G1
AA44196-G1EA 968 USD
AA44196-G1
Platinum ≥99.9% (metals basis), wire, Diameter 0.01 mm (0.0004 in)
Platinum

MDL: MFCD00011179


Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈1.68mg/m, Temper: hard Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR