Praseodymium ≥99.5% (REO, rare earth oxide basis), powder -40 mesh

Supplier: Thermo Scientific Chemicals
Danger

044183-06
AA44183-06EA 165 USD
AA44183-06
Praseodymium ≥99.5% (REO, rare earth oxide basis), powder -40 mesh
Praseodymium

Purity: 99.5% CAS No.: 7440-10-0 Physical Form: Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Grey to silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00011174

Formula: Pr
MW: 140.91
Storage Temperature: Ambient
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR