Lanthanum ≥99.7% (REO, rare earth oxide basis), powder -40 mesh

Supplier: Thermo Scientific Chemicals
Danger

044182-06 044182-14
AA44182-06EA 82.7 USD
AA44182-06 AA44182-14
Lanthanum ≥99.7% (REO, rare earth oxide basis), powder -40 mesh
Lanthanum

Purity: 99.7% CAS No.: 7439-91-0 Physical Form: Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Gray to silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00011066

Formula: La
Boiling Pt: 3464 °C (1013 hPa)
Melting Pt: 920 °C
Density: ∼6.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR