Aluminium ≥97.5% (metals basis), spherical powder, APS 4.5-7 µm
Supplier: Thermo Scientific Chemicals
|
Danger
|
044173-A7
044173-A1
AA44173-A7EA
471
USD
AA44173-A7
AA44173-A1
Aluminium ≥97.5% (metals basis), spherical powder, APS 4.5-7 µm
Aluminium
Tap density 1.5g/cm³
Purity: 97.5%
CAS No.: 7429-90-5
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Grey
Odor: Odorless
UN No.: 1396
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2057 °C (1013 hPa) Melting Pt: 659.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 UN: 1396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...