Gold ≥99.9% (metals basis), thin foil, Thickness 0.007 mm (0.00028 in)

Supplier: Thermo Scientific Chemicals

044139-FF 044139-FI
AA44139-FFEA 504 USD
AA44139-FF AA44139-FI
Gold ≥99.9% (metals basis), thin foil, Thickness 0.007 mm (0.00028 in)
Gold

MDL: MFCD00003436


Purity: 99.9% CAS No.: 7440-57-5 Physical Form: ≈0.08g/25x25mm, not certified pinhole free Odor: Odorless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR