Ethanol denatured, anhydrous ≥90% for HPLC, for LC-MS (denatured with 5% isopropanol and 5% methanol)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl alcohol denatured
Packaged under argon in resealable ChemSeal™ bottles.
044134-AE
AA44134-AEEA
53.06
USD
AA44134-AE
Ethanol denatured, anhydrous ≥90% for HPLC, for LC-MS (denatured with 5% isopropanol and 5% methanol)
Ethanol denatured
Purity: 90%
CAS No.: 64-17-5
Molecular Formula: C2H6O
Formula Weight: 46.08
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.789
Refractive Index: 1.3610
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Alcohol-like
Melting Point: -90°
Boiling Point: 78°
Beilstein Reference: 1718733
Merck Reference: 14,3760
UN No.: 1987
MDL No.: MFCD00003568
Formula:
H₃CCH₂OH MW: 46.07 g/mol Boiling Pt: 78 °C Melting Pt: –90 °C Density: 0.789 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1987 ADR: 3 II Merck Index: 14,3760 |
Specification Test Results
Evaporation residue | 5ppm max. |
H2O | 0.1% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...