Lithium-tert-butanolate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044133-04
044133-18
044133-09
AA44133-04EA
31.35
USD
AA44133-04
AA44133-18
AA44133-09
Lithium-tert-butanolate ≥99.9% (metals basis)
Lithium tert-butanolate
Purity: 99.9%
CAS No.: 1907-33-1
Molecular Formula: C4H9LiO
Formula Weight: 80.06
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White to cream
Odor: Odorless
Melting Point: ca 150° dec.
Beilstein Reference: 3620018
UN No.: 3206
MDL No.: MFCD00050479
Formula:
C₄H₉LiO Melting Pt: ∼ 150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00050479 CAS Number: 1907-33-1 EINECS: 217-611-5 UN: 3206 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...