Vanadium ≥99.8% (metals basis), wire, Diameter 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

044105-G1 044105-G5
AA44105-G1EA 80.1 USD
AA44105-G1 AA44105-G5
Vanadium ≥99.8% (metals basis), wire, Diameter 0.25 mm (0.01 in)
Vanadium

MDL: MFCD00011453


Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈0.3g/m Odor: Odorless MDL No.: MFCD00011453

Formula: V
Boiling Pt: 3477 °C (1013 hPa)
Melting Pt: 1896 °C
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR