Silver conductive adhesive paste

Supplier: Thermo Scientific Chemicals
Warning

044075-18
AA44075-18EA 629 USD
AA44075-18
Silver conductive adhesive paste
Silver conductive adhesive paste

Temperature stability range -65° to 275°C


Physical Form: Nominal electrical resistivity: 1x10-4ohm-cm Appearance: Silver paste Odor: Aromatic MDL No.: MFCD01868500

Storage Temperature: Ambient
MDL Number: MFCD01868500
EINECS: 232-033-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR