Ammonium tetrafluoroborate ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate
044055-22
044055-06
044055-14
AA44055-22EA
311
USD
AA44055-22
AA44055-06
AA44055-14
Ammonium tetrafluoroborate ≥99.999% (metals basis)
Ammonium tetrafluoroborate
MDL: MFCD06245785
Purity: 99.999%
CAS No.: 13826-83-0
Molecular Formula: NH4BF4
Formula Weight: 104.84
Physical Form: Crystalline
Density: 1.871
Appearance: White
Odor: Ammonia-like
Melting Point: 220° subl.
UN No.: 3260
MDL No.: MFCD06245785
Formula:
H₄BF₄N Melting Pt: 220 °C (subl.) Density: 1.871 Storage Temperature: Ambient |
MDL Number:
MFCD06245785 CAS Number: 13826-83-0 EINECS: 237-531-4 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...