Ammonium tetrafluoroborate ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

044055-22 044055-06 044055-14
AA44055-22EA 311 USD
AA44055-22 AA44055-06 AA44055-14
Ammonium tetrafluoroborate ≥99.999% (metals basis)
Ammonium tetrafluoroborate

MDL: MFCD06245785


Purity: 99.999% CAS No.: 13826-83-0 Molecular Formula: NH4BF4 Formula Weight: 104.84 Physical Form: Crystalline Density: 1.871 Appearance: White Odor: Ammonia-like Melting Point: 220° subl. UN No.: 3260 MDL No.: MFCD06245785

Formula: H₄BF₄N
Melting Pt: 220 °C (subl.)
Density: 1.871
Storage Temperature: Ambient
MDL Number: MFCD06245785
CAS Number: 13826-83-0
EINECS: 237-531-4
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR