Holmium ≥99.9% (metals basis), rods, Diameter 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
044050-HF
044050-DJ
AA44050-HFEA
320
USD
AA44050-HF
AA44050-DJ
Holmium ≥99.9% (metals basis), rods, Diameter 6.35 mm (0.25 in)
Holmium
MDL: MFCD00011049
Purity: 99.9%
CAS No.: 7440-60-0
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.28g/mm, Cast rod, diameter is approximate
Odor: Odorless
Formula:
Ho Storage Temperature: Ambient |
MDL Number:
MFCD00011049 CAS Number: 7440-60-0 EINECS: 231-169-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...