Lead ≥99.9% (metals basis), wire, Diameter 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Danger

044016-H2 044016-G6
AA44016-H2EA 461 USD
AA44016-H2 AA44016-G6
Lead ≥99.9% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Lead

MDL: MFCD00134050


Purity: 99.9% CAS No.: 7439-92-1 Physical Form: ≈35.6g/m Appearance: Grey Odor: Odorless MDL No.: MFCD00134050

Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740
Melting Pt: 327
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now

Specification Test Results

Form ~ 35.6g/m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR