Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), Co(TMHD)3
043993-09
043993-02
043993-04
AA43993-09EA
651
USD
AA43993-09
AA43993-02
AA43993-04
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III)
Applications: Vapor deposition
Purity: 99%
CAS No.: 14877-41-9
Molecular Formula: C33H57CoO6
Formula Weight: 608.72
Physical Form: Powder
Appearance: Green
Melting Point: 254-256°
Boiling Point: subl. 120°/0.5mm
MDL No.: MFCD00233617
Vapor deposition
Formula:
C₃₃H₅₇CoO₆ Boiling Pt: 120 °C (0.5 mmHg) (subl.) Melting Pt: 254…256 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233617 CAS Number: 14877-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...