Germanium ≥99.999% (metals basis), pieces ≤ 3.2 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

043986-09 043986-18
AA43986-09EA 219 USD
AA43986-09 AA43986-18
Germanium ≥99.999% (metals basis), pieces ≤ 3.2 mm (0.125 in)
Germanium

MDL: MFCD00085310


Purity: 99.999% CAS No.: 7440-56-4 Odor: Odorless MDL No.: MFCD00085310

Formula: Ge
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937.2 °C
Density: 5.323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR