Tetraammineplatinum(II) nitrate (≥50% Pt) ≥99.99%, Premion®

Supplier: Thermo Scientific Chemicals
Danger

043897-01 043897-06 043897-03
AA43897-01EA 95.8 USD
AA43897-01 AA43897-06 AA43897-03
Tetraammineplatinum(II) nitrate (≥50% Pt) ≥99.99%, Premion®
Tetraammineplatinum(II) nitrate

Purity: 99.99% CAS No.: 20634-12-2 Molecular Formula: H12N6O6Pt Formula Weight: 387.21 Physical Form: Crystalline Appearance: White Melting Point: 262° dec. UN No.: 1477 MDL No.: MFCD00011624

Formula: H₁₂N₆O₆Pt
Melting Pt: 262 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011624
CAS Number: 20634-12-2
EINECS: 243-929-9
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR