Silver, Oil Based Standard Solution, Specpure®, Ag 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
043868-18
AA43868-18EA
72.08
USD
AA43868-18
Silver, Oil Based Standard Solution, Specpure®, Ag 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Silver Standards
Physical Form: Liquid
MDL No.: MFCD00003397
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...