Silicon, Oil Based Standard Solution, Specpure®, Si 5000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
043867-18
AA43867-18EA
93.84
USD
AA43867-18
Silicon, Oil Based Standard Solution, Specpure®, Si 5000 µg/g, Thermo Scientific
Standards
Cationic Standards
Silicium Standards
Physical Form: Liquid
MDL No.: MFCD00085311
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...