Graphite, plate, highly oriented pyrolytic graphite (HOPG) 10x10x(1.6 min) mm (0.394x0.394x0.079 in)

Supplier: Thermo Scientific Chemicals

043837-KF
AA43837-KFEA 976 USD
AA43837-KF
Graphite, plate, highly oriented pyrolytic graphite (HOPG) 10x10x(1.6 min) mm (0.394x0.394x0.079 in)
Graphite

Physical Form: 0.4 +/-0.1° mosaic angle MDL No.: MFCD00144065

Formula: C
Melting Pt: 3827 °C
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR