Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness 0.15 mm (0.006 in)
Supplier: Thermo Scientific Chemicals
043813-FI
AA43813-FIEA
1517
USD
AA43813-FI
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness 0.15 mm (0.006 in)
Platinum Iridium alloy (80:20 w%)
MDL: MFCD02091683
Purity: 99.9%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈8.13g/50x50mm
Odor: Odorless
MDL No.: MFCD02091683
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...