Sulfur ≥99.5%, powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

043766-36 043766-A3
AA43766-36EA 69 USD
AA43766-36 AA43766-A3
Sulfur ≥99.5%, powder -325 mesh
Sulphur

Purity: 99.5% CAS No.: 7704-34-9 Appearance: Yellow Odor: Weak UN No.: 1350 MDL No.: MFCD00085316

Formula: S
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 113…119 °C
Density: 2.36 g/cm³ (20 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR